L-Arginin-methylester-dihydrochlorid 98%

Lieferant: Apollo Scientific

Synonyme: H-Arg-OMe·2HCL, H-Arg-OMe.2HCl

OR471616-100G
APOSOR471616-100GEA 112 EUR
APOSOR471616-100G
L-Arginin-methylester-dihydrochlorid 98%
L-Arginine methyl ester dihydrochloride
Formel: C₇H₁₆N₄O₂
Molecular Weight: 261,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00038948
CAS-Nummer: 26340-89-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR