L-Glutamic acid dibenzyl ester hydrochloride 97%

Lieferant: Apollo Scientific

OR471621-100G
APOSOR471621-100GEA 120 EUR
APOSOR471621-100G
L-Glutamic acid dibenzyl ester hydrochloride 97%
L-Glutamic acid dibenzyl ester hydrochloride
Formel: C₁₉H₂₁NO₄·HCl
Lagertemperatur: Raumtemperatur
MDL: MFCD00035103
CAS-Nummer: 4561-10-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR