H-Orn(boc)-OH 97%

Lieferant: Apollo Scientific

Synonyme: H-Orn(Boc)-OH, N-α-t-Boc-L-Ornithine, Boc-L-Orn-OH, Ndelta-Boc-L-ornithine, (S)-2-Amino-5-((tert-butoxycarbonyl)amino)pentanoic acid

OR471625-25G
APOSOR471625-25GEA 122 EUR
APOSOR471625-25G
H-Orn(boc)-OH 97%
H-Orn(boc)-OH
Formel: C₁₀H₂₀N₂O₄
Molecular Weight: 232.28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00037219
CAS-Nummer: 13650-49-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR