N-Boc-D-alanine methyl ester 98%
Lieferant: Apollo Scientific
Synonyme:
Boc-D-Ala-OMe, BOC-D-Alanine methyl ester, (R)-Methyl 2-((tert-butoxycarbonyl)amino)propanoate, BOC-D-Alanine methyl este
OR471627-25G
OR471627-100G
APOSOR471627-25GEA
27.7
EUR
APOSOR471627-25G
APOSOR471627-100G
N-Boc-D-alanine methyl ester 98%
N-Boc-D-alanine methyl ester
Formel:
C₉H₁₇NO₄ Molecular Weight: 203.238 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191865 CAS-Nummer: 91103-47-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...