Ethyl benzimidate hydrochloride 97%
Lieferant: Apollo Scientific
OR471631-100G
APOSOR471631-100GEA
0
EUR
APOSOR471631-100G
Ethyl benzimidate hydrochloride 97%
Ethyl benzimidate hydrochloride
Formel:
C₉H₁₂ClNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043246 CAS-Nummer: 5333-86-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...