Fmoc-Cys(tBu)-OH 98%

Lieferant: Apollo Scientific

Synonyme: N-Fmoc-S-(tert-butyl)-L-cysteine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(tert-butyl)-L-cysteine, N-Fmoc-S-tert-butyl-L-cysteine, Fmoc-Cys(tBu)-OH

OR471634-100G OR471634-25G
APOSOR471634-100GEA 209 EUR
APOSOR471634-100G APOSOR471634-25G
Fmoc-Cys(tBu)-OH 98%
Fmoc-Cys(tBu)-OH
Formel: C₂₂H₂₅NO₄S
Molecular Weight: 399.505 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00037130
CAS-Nummer: 67436-13-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR