4-methoxy-1-naphthoic acid 98%
Lieferant: Apollo Scientific
Synonyme:
4-Methoxynaphthalene-1-carboxylic acid, 4-Methoxy-1-naphthoic acid
OR471638-25G
APOSOR471638-25GEA
60.9
EUR
APOSOR471638-25G
4-methoxy-1-naphthoic acid 98%
4-methoxy-1-naphthoic acid
Formel:
C₁₂H₁₀O₃ Molecular Weight: 202,209 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02575372 CAS-Nummer: 13041-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...