(S)-3-[4-(5-Bromo-2-chlorobenzyl)phenoxy]tetrahydrofuran 97%

Lieferant: Apollo Scientific

Synonyme: (S)-3-(4-(5-Bromo-2-chlorobenzyl)phenoxy)tetrahydrofuran

OR471641-25G
APOSOR471641-25GEA 174 EUR
APOSOR471641-25G
(S)-3-[4-(5-Bromo-2-chlorobenzyl)phenoxy]tetrahydrofuran 97%
(S)-3-[4-(5-Bromo-2-chlorobenzyl)phenoxy]tetrahydrofuran
Formel: C₁₇H₁₆BrClO₂
Molecular Weight: 367.665 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27920793
CAS-Nummer: 915095-89-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR