N-Benzyloxycarbonyl-DL-Serin 98%
Lieferant: Apollo Scientific
Synonyme:
N-Cbz-DL-serine, Cbz-DL-Ser-OH, Z-DL-Ser-OH
OR471643-100G
APOSOR471643-100GEA
270
EUR
APOSOR471643-100G
N-Benzyloxycarbonyl-DL-Serin 98%
N-Benzyloxycarbonyl-DL-Serin
Formel:
C₁₁H₁₃NO₅ Molecular Weight: 239,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063143 CAS-Nummer: 2768-56-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...