(S)-β-(Carbobenzoxyamino)-γ-butyrolactone 98%

Lieferant: Apollo Scientific

Synonyme: (S)-Benzyl (5-oxotetrahydrofuran-3-yl)carbamate, Benzyl (S)-Tetrahydro-5-oxo-3-furanylcarbamate, (S)-4-(Carbobenzoxyamino)tetrahydrofuran-2-one, (S)-beta-(Cbz-amino)-gamma-butyrolactone, (S)-4-(Cbz-amino)tetrahydrofuran-2-one

OR471646-25G
APOSOR471646-25GEA 161 EUR
APOSOR471646-25G
(S)-β-(Carbobenzoxyamino)-γ-butyrolactone 98%
(S)-beta-(Carbobenzoxyamino)-gamma-butyrolactone
Formel: C₁₂H₁₃NO₄
Molecular Weight: 235.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00216569
CAS-Nummer: 87219-29-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR