4-[(2-Tetrahydropyranyl)oxy]phenol 98%

Lieferant: Apollo Scientific

Synonyme: Deoxyarbutin, 4-((Tetrahydro-2H-pyran-2-yl)oxy)phenol, 4-(Tetrahydropyran-2-yloxy)phenol

OR471648-100G
APOSOR471648-100GEA 142 EUR
APOSOR471648-100G
4-[(2-Tetrahydropyranyl)oxy]phenol 98%
4-[(2-Tetrahydropyranyl)oxy]phenol
Formel: C₁₁H₁₄O₃
Molecular Weight: 194,227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09970981
CAS-Nummer: 53936-56-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR