Fmoc-Asp-OBzl 95%

Lieferant: Apollo Scientific

Synonyme: N-Fmoc-L-aspartic acid 1-benzyl ester, Fmoc-Asp-Obzl

OR471652-10G OR471652-5G
APOSOR471652-10GEA 137 EUR
APOSOR471652-10G APOSOR471652-5G
Fmoc-Asp-OBzl 95%
Fmoc-Asp-OBzl
Formel: C₂₆H₂₃NO₆
Lagertemperatur: Raumtemperatur
MDL: MFCD00198201
CAS-Nummer: 86060-83-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR