Benzyl-(S)-(-)-lactate 98%

Lieferant: Apollo Scientific

Synonyme: Benzyl L-lactate, (S)-(-)-Lactic Acid Benzyl Ester, Benzyl (S)-Lactate

OR471654-25G
APOSOR471654-25GEA 151 EUR
APOSOR471654-25G
Benzyl-(S)-(-)-lactate 98%
Benzyl-(S)-(-)-lactate
Formel: C₁₀H₁₂O₃
Molecular Weight: 180.20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00209546
CAS-Nummer: 56777-24-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR