(R)-2-Hydroxy-2-(4-methoxyphenyl)acetic acid 95%

Lieferant: Apollo Scientific

OR471655-5G
APOSOR471655-5GEA 122 EUR
APOSOR471655-5G
(R)-2-Hydroxy-2-(4-methoxyphenyl)acetic acid 95%
(R)-2-Hydroxy-2-(4-methoxyphenyl)acetic acid
Formel: C₉H₁₀O₄
Molecular Weight: 182.1733 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00798434
CAS-Nummer: 20714-89-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR