(S)-N-Boc-3-Amino-3-phenyl-1-propanol 97%

Lieferant: Apollo Scientific

OR471656-5G
APOSOR471656-5GEA 0 EUR
APOSOR471656-5G
(S)-N-Boc-3-Amino-3-phenyl-1-propanol 97%
(S)-N-Boc-3-Amino-3-phenyl-1-propanol
Formel: C₁₄H₂₁NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09025325
CAS-Nummer: 718611-17-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR