2-Phenylpyridin-4-amine 97%

Lieferant: Apollo Scientific

Synonyme: 4-Amino-2-phenylpyridine

OR471657-5G
APOSOR471657-5GEA 224 EUR
APOSOR471657-5G
2-Phenylpyridin-4-amine 97%
2-Phenylpyridin-4-amine
Formel: C₁₁H₁₀N₂
Molecular Weight: 170.2100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01646280
CAS-Nummer: 21203-86-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR