6-Methyl-1H-indole-3-carboxylic acid 98%

Lieferant: Apollo Scientific

OR471658-1G OR471658-5G
APOSOR471658-1GEA 65.4 EUR
APOSOR471658-1G APOSOR471658-5G
6-Methyl-1H-indole-3-carboxylic acid 98%
6-Methyl-1H-indole-3-carboxylic acid
Formel: C₁₀H₉NO₂
Molecular Weight: 175.1840 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06203633
CAS-Nummer: 209920-43-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR