(6-Methoxy-3-pyridyl)methanol 97%

Lieferant: Apollo Scientific

OR471668-5G
APOSOR471668-5GEA 82.8 EUR
APOSOR471668-5G
(6-Methoxy-3-pyridyl)methanol 97%
(6-Methoxy-3-pyridyl)methanol
Formel: C₇H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06202826
CAS-Nummer: 58584-63-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR