6-(Chloromethyl)pyridine-2-carbonitrile 95%

Lieferant: Apollo Scientific

OR471675-5G
APOSOR471675-5GEA 0 EUR
APOSOR471675-5G
6-(Chloromethyl)pyridine-2-carbonitrile 95%
6-(Chloromethyl)pyridine-2-carbonitrile
Formel: C₇H₅ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09991809
CAS-Nummer: 135450-23-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR