(±)-1,2,3,4-Tetrahydro-2-naphthol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,2,3,4-Tetrahydronaphthalen-2-ol

OR471677-1G
APOSOR471677-1GEA 43.5 EUR
APOSOR471677-1G
(±)-1,2,3,4-Tetrahydro-2-naphthol 98%
(±)-1,2,3,4-Tetrahydro-2-naphthol
Formel: C₁₀H₁₂O
Molecular Weight: 148,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00045575
CAS-Nummer: 530-91-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR