Fmoc-4-cyano-D-phenylalanine 95%

Lieferant: Apollo Scientific

Synonyme: Fmoc-D-Phe(4-CN)-OH

OR471678-5G
APOSOR471678-5GEA 78.3 EUR
APOSOR471678-5G
Fmoc-4-cyano-D-phenylalanine 95%
Fmoc-4-cyano-D-phenylalanine
Formel: C₂₅H₂₀N₂O₄
Molecular Weight: 412.45 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00672560
CAS-Nummer: 205526-34-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR