Boc-D-2,4-diaminobutyric acid 97%

Lieferant: Apollo Scientific

Synonyme: Boc-D-Dab-OH

OR471679-5G
APOSOR471679-5GEA 244 EUR
APOSOR471679-5G
Boc-D-2,4-diaminobutyric acid 97%
Boc-D-2,4-diaminobutyric acid
Formel: C₉H₁₈N₂O₄
Molecular Weight: 218.253 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00798622
CAS-Nummer: 80445-78-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR