4-Bromo-1H-indazole-6-carboxylic acid 97%

Lieferant: Apollo Scientific

OR471681-1G
APOSOR471681-1GEA 111 EUR
APOSOR471681-1G
4-Bromo-1H-indazole-6-carboxylic acid 97%
4-Bromo-1H-indazole-6-carboxylic acid
Formel: C₈H₅BrN₂O₂
Molecular Weight: 241.0415 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07781800
CAS-Nummer: 885523-43-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR