2-Amino-5-[(4-ethyl-1-piperazinyl)methyl]pyridine 95%
Lieferant: Apollo Scientific
Synonyme:
5-((4-Ethylpiperazin-1-yl)methyl)pyridin-2-amine
OR471684-25G
OR471684-5G
APOSOR471684-25GEA
244
EUR
APOSOR471684-25G
APOSOR471684-5G
2-Amino-5-[(4-ethyl-1-piperazinyl)methyl]pyridine 95%
2-Amino-5-[(4-ethyl-1-piperazinyl)methyl]pyridine
Formel:
C₁₂H₂₀N₄ Molecular Weight: 220.314 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14706690 CAS-Nummer: 1180132-17-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...