2-Chlor-5-methylnicotinsäure 98%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-5-methylpyridine-3-carboxylic acid

OR471690-5G
APOSOR471690-5GEA 66.7 EUR
APOSOR471690-5G
2-Chlor-5-methylnicotinsäure 98%
2-Chlor-5-methylnicotinsäure
Formel: C₇H₆ClNO₂
Molecular Weight: 171.5800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07375371
CAS-Nummer: 66909-30-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR