Methyl-3-brom-1H-indazol-5-carboxylat 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 3-bromo-1H-indazole-5-carboxylate
OR471693-5G
APOSOR471693-5GEA
136
EUR
APOSOR471693-5G
Methyl-3-brom-1H-indazol-5-carboxylat 95%
Methyl-3-brom-1H-indazol-5-carboxylat
Formel:
C₉H₇BrN₂O₂ Molecular Weight: 255.0681 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11045401 CAS-Nummer: 1086391-06-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...