8-Azabicyclo[3.2.1]octan-3-onhydrochlorid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 8-Azabicyclo[3.2.1]octan-3-one hydrochloride

OR4717-5G
APOSOR4717-5GEA 21.9 EUR
APOSOR4717-5G
8-Azabicyclo[3.2.1]octan-3-onhydrochlorid 97%
Nortropinone hydrochloride
Formel: C₇H₁₂ClNO
Molecular Weight: 161,63 g/mol
Schmelzpunkt: 204 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03613582
CAS-Nummer: 25602-68-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR