2-Aminobenzophenon 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: o-Aminobenzophenon

OR4724-5G
APOSOR4724-5GEA 21.9 EUR
APOSOR4724-5G
2-Aminobenzophenon 95%
2-Aminobenzophenon
Formel: C₁₃H₁₁NO
Molecular Weight: 197,24 g/mol
Schmelzpunkt: 103…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007713
CAS-Nummer: 2835-77-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR