4-Bromopyridine-2,6-dicarboxamide

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,6-Bis(aminocarbonyl)-4-bromopyridine, 4-Bromo-2,6-dicarbamoylpyridine

OR4731-250MG
APOSOR4731-250MGEA 154 EUR
APOSOR4731-250MG
4-Bromopyridine-2,6-dicarboxamide
4-Bromopyridine-2,6-dicarboxamide
Formel: C₇H₆BrN₃O₂
Molecular Weight: 244,045 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04113814
CAS-Nummer: 668992-72-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR