2-(2,6-Dichlorophenyl)ethanimidamide hydrochloride 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,6-Dichlorophenylacetamidine hydrochloride

OR4732-1G
APOSOR4732-1GEA 72.5 EUR
APOSOR4732-1G
2-(2,6-Dichlorophenyl)ethanimidamide hydrochloride 98%
2-(2,6-Dichlorophenyl)ethanimidamide hydrochloride
Formel: C₈H₉Cl₃N₂
Molecular Weight: 239,53 g/mol
Schmelzpunkt: 323 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00053014
CAS-Nummer: 175276-76-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR