3-Bromo-2,6-diaminopyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Bromopyridine-2,6-diamine
OR4741-500MG
APOSOR4741-500MGEA
21.9
EUR
APOSOR4741-500MG
3-Bromo-2,6-diaminopyridine
3-Bromo-2,6-diaminopyridine
Formel:
C₅H₆BrN₃ Molecular Weight: 188.03 g/mol Schmelzpunkt: 171…176 °C Dichte: 1.05…1.20 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD04113819 CAS-Nummer: 54903-86-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...