(4-Acetylphenyl)phenylmethane

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Benzylphenyl)ethan-1-one

OR4747-1G
APOSOR4747-1GEA 40.6 EUR
APOSOR4747-1G
(4-Acetylphenyl)phenylmethane
(4-Acetylphenyl)phenylmethane
Formel: C₁₅H₁₄O
Molecular Weight: 210,28 g/mol
Schmelzpunkt: 38…40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00079700
CAS-Nummer: 782-92-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR