(1R)-1-(3-Bromophenyl)-N-methyl-ethanamine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR475155-1G OR475155-250MG
APOSOR475155-1GEA 575 EUR
APOSOR475155-1G APOSOR475155-250MG
(1R)-1-(3-Bromophenyl)-N-methyl-ethanamine hydrochloride
(1R)-1-(3-Bromophenyl)-N-methyl-ethanamine hydrochloride
Formel: C₉H₁₂BrN·ClH
Molecular Weight: 250.563 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31714839
CAS-Nummer: 2366997-08-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR