2-Brom-3,5-dichlorpyridin
Lieferant: Apollo Scientific
|
Achtung
|
OR4756-1G
APOSOR4756-1GEA
21.9
EUR
APOSOR4756-1G
2-Brom-3,5-dichlorpyridin
2-Bromo-3,5-dichloropyridine
Formel:
C₅H₂BrCl₂N Schmelzpunkt: 43 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00233990 CAS-Nummer: 14482-51-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...