6-Hydroxy-3-pyridinesulphonic acid

Lieferant: Apollo Scientific

OR4767-1G
APOSOR4767-1GEA 0 EUR
APOSOR4767-1G
6-Hydroxy-3-pyridinesulphonic acid
6-Hydroxy-3-pyridinesulphonic acid
Formel: C₅H₅NO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD01574841
CAS-Nummer: 6684-46-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR