4-Amino-6-(tert-butyl)pyrimidine
Lieferant: Apollo Scientific
|
Achtung
|
OR4772-250MG
APOSOR4772-250MGEA
58
EUR
APOSOR4772-250MG
4-Amino-6-(tert-butyl)pyrimidine
4-Amino-6-(tert-butyl)pyrimidine
Formel:
C₈H₁₃N₃ Schmelzpunkt: 170…171 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234463 CAS-Nummer: 3435-27-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...