5-Bromo-2-furoic acid hydrazide

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromofuran-2-carbohydrazide

OR4776-1G
APOSOR4776-1GEA 24.8 EUR
APOSOR4776-1G
5-Bromo-2-furoic acid hydrazide
5-Bromo-2-furoic acid hydrazide
Formel: C₅H₅BrN₂O₂
Molecular Weight: 205,02 g/mol
Schmelzpunkt: 136…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01310787
CAS-Nummer: 89282-37-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR