4,6-Dimethylnicotinic acid hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,6-Dimethylnicotinic acid

OR4777-100MG
APOSOR4777-100MGEA 21.9 EUR
APOSOR4777-100MG
4,6-Dimethylnicotinic acid hydrochloride
4,6-Dimethylnicotinic acid hydrochloride
Formel: C₈H₁₀ClNO₂
Molecular Weight: 187,63 g/mol
Schmelzpunkt: 193…194 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD19103652
CAS-Nummer: 22047-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR