3-Amino-2-benzo[b]thiophencarbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR4781-250MG
APOSOR4781-250MGEA
78.3
EUR
APOSOR4781-250MG
3-Amino-2-benzo[b]thiophencarbonsäure
3-Amino-2-benzo[b]thiophencarbonsäure
Formel:
C₉H₇NO₂S Lagertemperatur: Kühlschrank |
CAS-Nummer:
40142-71-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...