1-(tert-Butyl)piperazine dihydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR47826-250MG
APOSOR47826-250MGEA
27.7
EUR
APOSOR47826-250MG
1-(tert-Butyl)piperazine dihydrochloride
1-(tert-Butyl)piperazine dihydrochloride
Formel:
C₈H₁₈N₂·2ClH Molecular Weight: 215,164 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09759216 CAS-Nummer: 110469-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...