Ethyl 6-chloro-4-ethoxynicotinate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 6-chloro-4-ethoxypyridine-3-carboxylate, 2-Chloro-4-ethoxy-5-(ethoxycarbonyl)pyridine

OR47832-100MG
APOSOR47832-100MGEA 21.9 EUR
APOSOR47832-100MG
Ethyl 6-chloro-4-ethoxynicotinate
Ethyl 6-chloro-4-ethoxynicotinate
Formel: C₁₀H₁₂ClNO₃
Molecular Weight: 229,66 g/mol
Schmelzpunkt: 73…74 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09702463
CAS-Nummer: 40296-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR