Ethyl 3-amino-5-bromopyridine-2-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-5-bromo-2-(ethoxycarbonyl)pyridine, Ethyl 3-amino-5-bromopicolinate

OR47833-250MG OR47833-1G
APOSOR47833-250MGEA 149 EUR
APOSOR47833-250MG APOSOR47833-1G
Ethyl 3-amino-5-bromopyridine-2-carboxylate 95%
Ethyl 3-amino-5-bromopyridine-2-carboxylate
Formel: C₈H₉BrN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10566234
CAS-Nummer: 1334405-60-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR