2,6-Dichloro-4-(hydroxymethyl)benzaldehyde

Lieferant: Apollo Scientific
Achtung

OR47849-1G
APOSOR47849-1GEA 638 EUR
APOSOR47849-1G
2,6-Dichloro-4-(hydroxymethyl)benzaldehyde
2,6-Dichloro-4-(hydroxymethyl)benzaldehyde
Formel: C₈H₆Cl₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18904376
CAS-Nummer: 616195-96-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR