2,3-Dimethyl-4-methoxybenzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Methoxy-2,3-dimethylbenzoic acid

OR47850-1G
APOSOR47850-1GEA 72.5 EUR
APOSOR47850-1G
2,3-Dimethyl-4-methoxybenzoic acid
2,3-Dimethyl-4-methoxybenzoic acid
Formel: C₁₀H₁₂O₃
Molecular Weight: 180.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06203271
CAS-Nummer: 5628-61-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR