4-Bromobenzaldehyde oxime

Lieferant: Apollo Scientific
Achtung

OR47851-1G
APOSOR47851-1GEA 0 EUR
APOSOR47851-1G
4-Bromobenzaldehyde oxime
4-Bromobenzaldehyde oxime
Formel: C₇H₆BrNO
Lagertemperatur: Raumtemperatur
MDL: MFCD00463678
CAS-Nummer: 34158-73-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR