3-Amino-4-cyanobenzoic acid 97%

Lieferant: Apollo Scientific

OR47858-1G
APOSOR47858-1GEA 104 EUR
APOSOR47858-1G
3-Amino-4-cyanobenzoic acid 97%
3-Amino-4-cyanobenzoic acid
Formel: C₈H₆N₂O₂
Molecular Weight: 162.145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20640420
CAS-Nummer: 159847-71-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR