3-Amino-5-cyanobenzoic acid ≥97%

Lieferant: Apollo Scientific

OR47859-250MG
APOSOR47859-250MGEA 120 EUR
APOSOR47859-250MG
3-Amino-5-cyanobenzoic acid ≥97%
3-Amino-5-cyanobenzoic acid
Formel: C₈H₆N₂O₂
Molecular Weight: 162,145 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09263225
CAS-Nummer: 1000341-18-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR