N-Boc-3,4-dihydro-2H-pyridine

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 3,4-dihydropyridine-1(2H)-carboxylate

OR47889-1G
APOSOR47889-1GEA 112 EUR
APOSOR47889-1G
N-Boc-3,4-dihydro-2H-pyridine
N-Boc-3,4-dihydro-2H-pyridine
Formel: C₁₀H₁₇NO₂
Molecular Weight: 183,247 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00798175
CAS-Nummer: 131667-57-7
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR