3-(4-Aminophenyl)propionsäure 95%

Lieferant: Apollo Scientific

Synonyme: 3-(4-Aminophenyl)propansäure

OR4789-5G
APOSOR4789-5GEA 79.9 EUR
APOSOR4789-5G
3-(4-Aminophenyl)propionsäure 95%
3-(4-Aminophenyl)propionsäure
Formel: C₉H₁₁NO₂
Schmelzpunkt: 135…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017118
CAS-Nummer: 2393-17-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR