2-(3-Pyrrolidinyl)acetic acid hydrochloride
Lieferant: Apollo Scientific
Synonyme:
3-Pyrrolidineacetic acid
OR47890-1G
APOSOR47890-1GEA
284
EUR
APOSOR47890-1G
2-(3-Pyrrolidinyl)acetic acid hydrochloride
2-(3-Pyrrolidinyl)acetic acid hydrochloride
Formel:
C₆H₁₂ClNO₂ Molecular Weight: 165,62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06795694 CAS-Nummer: 89203-64-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...